Product Details

SNP ID
rs141796615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27433381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGTGTCACCTCCTGTGACCTC[C/T]ACAACCTCAGCTGCTTCCCCAGAGG
Phenotype
MIM: 606010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRTCAP3 PubMed Links

Gene Details

Gene
KRTCAP3
Gene Name
keratinocyte associated protein 3
There are no transcripts associated with this gene.

Gene
NRBP1
Gene Name
nuclear receptor binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321357.1 940 Silent Mutation TCC,TCT S36S NP_001308286.1
NM_001321358.1 940 Silent Mutation TCC,TCT S36S NP_001308287.1
NM_001321359.1 940 Silent Mutation TCC,TCT S36S NP_001308288.1
NM_001321361.1 940 Silent Mutation TCC,TCT S36S NP_001308290.1
NM_001321362.1 940 Silent Mutation TCC,TCT S36S NP_001308291.1
NM_001321363.1 940 Silent Mutation TCC,TCT S36S NP_001308292.1
NM_013392.3 940 Silent Mutation TCC,TCT S36S NP_037524.1

View Full Product Details