Product Details

SNP ID
rs141807943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222201428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTTACGCGATATCTGGCTTGA[A/G]ATAATGAAAGGCACCTGTAAGGAAA
Phenotype
MIM: 606597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAX3 PubMed Links

Gene Details

Gene
PAX3
Gene Name
paired box 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000438.5 1816 Intron NP_000429.2
NM_001127366.2 1816 Missense Mutation CTC,TTC L478F NP_001120838.1
NM_013942.4 1816 Intron NP_039230.1
NM_181457.3 1816 Intron NP_852122.1
NM_181458.3 1816 Missense Mutation CTC,TTC L479F NP_852123.1
NM_181459.3 1816 Missense Mutation CTC,TTC L479F NP_852124.1
NM_181460.3 1816 Silent Mutation ATC,ATT I396I NP_852125.1
NM_181461.3 1816 Silent Mutation ATC,ATT I396I NP_852126.1

View Full Product Details