Product Details
- SNP ID
-
rs141835303
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:219173018 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACGGGAGGTCAGTCCAAGGTCAGG[C/T]GGCCCAGGTGTAGGGATGCAGGACA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNPPD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1043160] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNPPD1
- Gene Name
- cyclin Pas1/PHO80 domain containing 1
- Gene
- FAM134A
- Gene Name
- family with sequence similarity 134 member A
There are no transcripts associated with this gene.
- Gene
- SLC23A3
- Gene Name
- solute carrier family 23 member 3
There are no transcripts associated with this gene.
View Full Product Details