Product Details

SNP ID
rs141976852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86216945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGTGAGGCTGCACACTCAAAGC[A/G]CACCCAGCTTGCGGATTTCTATGTT
Phenotype
MIM: 611841 MIM: 609139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL35 PubMed Links

Gene Details

Gene
MRPL35
Gene Name
mitochondrial ribosomal protein L35
There are no transcripts associated with this gene.

Gene
REEP1
Gene Name
receptor accessory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164730.1 1011 UTR 3 NP_001158202.1
NM_001164731.1 1011 UTR 3 NP_001158203.1
NM_001164732.1 1011 UTR 3 NP_001158204.1
NM_022912.2 1011 UTR 3 NP_075063.1
XM_005264502.1 1011 UTR 3 XP_005264559.1
XM_005264504.1 1011 UTR 3 XP_005264561.1
XM_011533044.1 1011 UTR 3 XP_011531346.1
XM_011533045.2 1011 UTR 3 XP_011531347.1
XM_017004725.1 1011 UTR 3 XP_016860214.1
XM_017004726.1 1011 UTR 3 XP_016860215.1
XM_017004727.1 1011 UTR 3 XP_016860216.1

View Full Product Details