Product Details

SNP ID
rs142015388
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174560656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTCTCTTTGAGTAAAAAGCCCA[A/G]ATGACTGTATGGAAGAGAGATGAAT
Phenotype
MIM: 602357
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
WIPF1 PubMed Links

Gene Details

Gene
WIPF1
Gene Name
WAS/WASL interacting protein family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077269.1 3567 UTR 3 NP_001070737.1
NM_003387.4 3567 UTR 3 NP_003378.3
XM_011511780.2 3567 UTR 3 XP_011510082.1

View Full Product Details