Product Details

SNP ID
rs142054687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113107426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTTAATATAAAATTGTGTACCT[A/G]TCAAAAATATCTTTTGGCCAGGCGT
Phenotype
MIM: 147679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL1RN PubMed Links

Gene Details

Gene
IL1RN
Gene Name
interleukin 1 receptor antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000577.4 Intron NP_000568.1
NM_001318914.1 Intron NP_001305843.1
NM_173841.2 Intron NP_776213.1
NM_173842.2 Intron NP_776214.1
NM_173843.2 Intron NP_776215.1
XM_005263661.4 Intron XP_005263718.1
XM_011511121.1 Intron XP_011509423.1

View Full Product Details