Product Details

SNP ID
rs142231194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99327594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTCACTCTCCTTGACTTCTTG[A/C]AAAAAGTCTCTTTCACTAGGGATTT
Phenotype
MIM: 606086 MIM: 612564
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EIF5B PubMed Links

Gene Details

Gene
EIF5B
Gene Name
eukaryotic translation initiation factor 5B
There are no transcripts associated with this gene.

Gene
TXNDC9
Gene Name
thioredoxin domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005783.3 399 Missense Mutation TTG,TTT L83F NP_005774.2
XM_017003147.1 399 Missense Mutation TTG,TTT L83F XP_016858636.1

View Full Product Details