Product Details

SNP ID
rs142486322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178465872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATATCTTCTAAAACTGCATCCTG[A/G]TATGACTTGTCACGTGGCTTCTCAT
Phenotype
MIM: 610219 MIM: 607062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DFNB59 PubMed Links

Gene Details

Gene
DFNB59
Gene Name
deafness, autosomal recessive 59
There are no transcripts associated with this gene.

Gene
FKBP7
Gene Name
FK506 binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135212.1 1812 Silent Mutation TAC,TAT Y188Y NP_001128684.1
NM_181342.2 1812 Silent Mutation TAC,TAT Y189Y NP_851939.1
XM_005246638.4 1812 UTR 3 XP_005246695.1
XM_011511348.2 1812 Silent Mutation TAC,TAT Y82Y XP_011509650.1
XM_011511349.2 1812 Silent Mutation TAC,TAT Y81Y XP_011509651.1

View Full Product Details