Product Details

SNP ID
rs142734461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127852979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAACGACTGAAACGGGCTGTG[C/T]AGTTTAATGTTTTCATGAAGACTTC
Phenotype
MIM: 606017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMMECR1L PubMed Links

Gene Details

Gene
AMMECR1L
Gene Name
AMMECR1 like
There are no transcripts associated with this gene.

Gene
LOC107985803
Gene Name
uncharacterized LOC107985803
There are no transcripts associated with this gene.

Gene
POLR2D
Gene Name
RNA polymerase II subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004805.3 255 Missense Mutation TAC,TGC Y67C NP_004796.1

View Full Product Details