Product Details

SNP ID
rs143232644
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240611356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGTTTGAACCTACCATCTGCT[A/G]TTTGTTTCCTATTCATCTCCTCTGT
Phenotype
MIM: 602646
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
GPR35 PubMed Links

Gene Details

Gene
GPR35
Gene Name
G protein-coupled receptor 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195381.1 Intron NP_001182310.1
NM_001195382.1 Intron NP_001182311.1
NM_005301.3 Intron NP_005292.2

View Full Product Details