Product Details
- SNP ID
-
rs143272788
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:60942395 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCTTTGTCTCCAAATTTGAAAG[C/T]TAGTCATCCAGAGCAGGTGGCCAGT
- Phenotype
-
MIM: 612787
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NONOP2
PubMed Links
Gene Details
- Gene
- NONOP2
- Gene Name
- non-POU domain containing, octamer-binding pseudogene 2
There are no transcripts associated with this gene.
- Gene
- PUS10
- Gene Name
- pseudouridylate synthase 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001322123.1 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
NP_001309052.1 |
NM_001322124.1 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
NP_001309053.1 |
NM_001322127.1 |
1729 |
Silent Mutation |
TAA,TAG |
*307* |
NP_001309056.1 |
NM_144709.3 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
NP_653310.2 |
XM_011532568.2 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530870.1 |
XM_011532570.1 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530872.1 |
XM_011532571.1 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530873.1 |
XM_011532572.1 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530874.1 |
XM_011532573.2 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530875.1 |
XM_011532574.2 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530876.1 |
XM_011532576.2 |
1729 |
Silent Mutation |
TAA,TAG |
*530* |
XP_011530878.1 |
XM_011532578.1 |
1729 |
Silent Mutation |
TAA,TAG |
*467* |
XP_011530880.1 |
XM_017003428.1 |
1729 |
Silent Mutation |
TAA,TAG |
*445* |
XP_016858917.1 |
XM_017003429.1 |
1729 |
Silent Mutation |
TAA,TAG |
*445* |
XP_016858918.1 |
View Full Product Details