Product Details

SNP ID
rs143323460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45006342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGACGGCAGCCCAGGGGGCGGC[A/G]GGCTGAGGAGCAGTGCGGGGCTGGA
Phenotype
MIM: 604994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIX2 PubMed Links

Gene Details

Gene
SIX2
Gene Name
SIX homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016932.4 997 Missense Mutation CCG,CTG P235L NP_058628.3
XM_005264100.3 997 Missense Mutation CCG,CTG P237L XP_005264157.1

View Full Product Details