Product Details

SNP ID
rs143388171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:60452581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGGCTTCAAATTTTCTCAGAA[C/T]TTAAGGGCTCTCGAGCTTCCATCCG
Phenotype
MIM: 606557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL11A PubMed Links

Gene Details

Gene
BCL11A
Gene Name
B-cell CLL/lymphoma 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018014.3 2696 Silent Mutation AAA,AAG K772K NP_060484.2
NM_022893.3 2696 Intron NP_075044.2
NM_138559.1 2696 Missense Mutation AAT,AGT N239S NP_612569.1
XM_011532909.1 2696 UTR 3 XP_011531211.1
XM_011532910.1 2696 Missense Mutation AAT,AGT N823S XP_011531212.1
XM_011532912.1 2696 Missense Mutation AAT,AGT N789S XP_011531214.1
XM_017004333.1 2696 Intron XP_016859822.1
XM_017004334.1 2696 Intron XP_016859823.1
XM_017004335.1 2696 Intron XP_016859824.1
XM_017004336.1 2696 Intron XP_016859825.1
XM_017004337.1 2696 Intron XP_016859826.1
XM_017004338.1 2696 Intron XP_016859827.1

View Full Product Details