Product Details

SNP ID
rs143710089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:210293772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTCTTGACCTGCCATCAGGGC[C/T]TCCACTTCTTCCTCTTTCATCTTTT
Phenotype
MIM: 160780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYL1 PubMed Links

Gene Details

Gene
MYL1
Gene Name
myosin light chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_079420.2 446 Silent Mutation GAA,GAG E169E NP_524144.1
NM_079422.2 446 Silent Mutation GAA,GAG E125E NP_524146.1

View Full Product Details