Product Details

SNP ID
rs143800390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85579138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGGGGAAAACTTGGAACATCTC[C/T]GCAACAAGACAGAGGATCTGGAAGC
Phenotype
MIM: 607029 MIM: 603177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VAMP5 PubMed Links

Gene Details

Gene
VAMP5
Gene Name
vesicle associated membrane protein 5
There are no transcripts associated with this gene.

Gene
VAMP8
Gene Name
vesicle associated membrane protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003761.4 245 Missense Mutation CGC,TGC R45C NP_003752.2
XM_017005170.1 245 Missense Mutation CGC,TGC R45C XP_016860659.1

View Full Product Details