Product Details
- SNP ID
-
rs143927979
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:202035874 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTAGACGGGGACCTGCTGAGCGG[A/G]GTGTGCTACGTTGGCCTCTCCAGTG
- Phenotype
-
MIM: 603410
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FZD7
PubMed Links
Gene Details
- Gene
- FZD7
- Gene Name
- frizzled class receptor 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003507.1 |
1288 |
Silent Mutation |
GGA,GGG |
G409G |
NP_003498.1 |
View Full Product Details