Product Details

SNP ID
rs144200621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74959382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTGCAAAAGATGCCTACTGTTG[C/T]GCTCAGCAGGGAAAAAGGAAAACCC
Phenotype
MIM: 607269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105374809 PubMed Links

Gene Details

Gene
LOC105374809
Gene Name
uncharacterized LOC105374809
There are no transcripts associated with this gene.

Gene
POLE4
Gene Name
DNA polymerase epsilon 4, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019896.2 287 Silent Mutation TGC,TGT C85C NP_063949.2

View Full Product Details