Product Details

SNP ID
rs144260473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227815483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAACTTTGACTGCTGTCTTGGA[C/T]ACACAGACCGTATTCTTCATCCTAA
Phenotype
MIM: 601960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL20 PubMed Links

Gene Details

Gene
CCL20
Gene Name
C-C motif chemokine ligand 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130046.1 173 Missense Mutation CAC,TAC H35Y NP_001123518.1
NM_004591.2 173 Missense Mutation CAC,TAC H36Y NP_004582.1

View Full Product Details