Product Details

SNP ID
rs144332302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26356065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGCCAGCATTCCTGGCCTCAAG[C/T]CTTGTATTGATTAAAAGTACATTAA
Phenotype
MIM: 607915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRF3 PubMed Links

Gene Details

Gene
ADGRF3
Gene Name
adhesion G protein-coupled receptor F3
There are no transcripts associated with this gene.

Gene
EPT1
Gene Name
ethanolaminephosphotransferase 1
There are no transcripts associated with this gene.

View Full Product Details