Product Details

SNP ID
rs144433180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112831327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGGGCTTATCATCTTTCAACA[C/T]GCAGGACAGGTACAGATTCTTTTCC
Phenotype
MIM: 147720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1B PubMed Links

Gene Details

Gene
IL1B
Gene Name
interleukin 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000576.2 649 Missense Mutation CTG,TTG L188L NP_000567.1
XM_017003988.1 649 Missense Mutation CTG,TTG L157L XP_016859477.1

View Full Product Details