Product Details

SNP ID
rs144459824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27215502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAAGCAAACCCCCTCAAAGGT[A/G]ACTTGGCCAACACCTTCCGTGGCTT
Phenotype
MIM: 114010 MIM: 604024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRAID PubMed Links

Gene Details

Gene
ATRAID
Gene Name
all-trans retinoic acid induced differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170795.1 660 Missense Mutation AAC,GAC N108D NP_001164266.1
NM_016085.4 660 Missense Mutation AAC,GAC N50D NP_057169.2
NM_080592.3 660 Missense Mutation AAC,GAC N163D NP_542159.3
Gene
CAD
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
There are no transcripts associated with this gene.

Gene
SLC5A6
Gene Name
solute carrier family 5 member 6
There are no transcripts associated with this gene.

View Full Product Details