Product Details

SNP ID
rs144464562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108378349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTCTTACACTAATGGCCTTACA[C/T]GACATGGAGGATTTTACATTTGATG
Phenotype
MIM: 608357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SULT1C4 PubMed Links

Gene Details

Gene
SULT1C4
Gene Name
sulfotransferase family 1C member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321770.1 439 Silent Mutation CAC,CAT H4H NP_001308699.1
NM_006588.3 439 Silent Mutation CAC,CAT H4H NP_006579.2
XM_017003807.1 439 Intron XP_016859296.1

View Full Product Details