Product Details

SNP ID
rs144469983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68866194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCACTGGTATCTATTTCTAGCC[A/G]TCCACTGCTGGCATCCTCAGCTTCA
Phenotype
MIM: 608748
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP10 PubMed Links

Gene Details

Gene
BMP10
Gene Name
bone morphogenetic protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014482.1 871 Missense Mutation NP_055297.1

View Full Product Details