Product Details

SNP ID
rs144577936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79122056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCCAGGCCGAGGGTGCCTTTGT[A/G]GCCTCACTGATTAAGGAGAGTGGCA
Phenotype
MIM: 167770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
REG1A PubMed Links

Gene Details

Gene
REG1A
Gene Name
regenerating family member 1 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002909.4 355 Silent Mutation GTA,GTG V84V NP_002900.2

View Full Product Details