Product Details

SNP ID
rs144647588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161316481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACTTGGGGAAGTTGTACCTGC[C/T]AGAATTAGCAAGAGCTTTCTTTAAG
Phenotype
MIM: 607173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMD14 PubMed Links

Gene Details

Gene
PSMD14
Gene Name
proteasome 26S subunit, non-ATPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005805.5 375 UTR 5 NP_005796.1

View Full Product Details