Product Details

SNP ID
rs144727732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219574888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCTCTGAGCCCCTGCTAGGCCTG[C/T]TGGCACTGTCACCGGGAGGACCCGT
Phenotype
MIM: 147380 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INHA PubMed Links

Gene Details

Gene
INHA
Gene Name
inhibin alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002191.3 606 Silent Mutation CTG,TTG L155L NP_002182.1
Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

View Full Product Details