Product Details

SNP ID
rs145311492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218823859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAAGAGATGCTGGGTCTCGTCC[A/G]CTAGCACCAGCCTGTGTACCTGTGG
Phenotype
MIM: 606530 MIM: 604976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP27A1 PubMed Links

Gene Details

Gene
CYP27A1
Gene Name
cytochrome P450 family 27 subfamily A member 1
There are no transcripts associated with this gene.

Gene
MIR9500
Gene Name
microRNA 9500
There are no transcripts associated with this gene.

Gene
PRKAG3
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017431.2 1392 Missense Mutation GCG,GTG A458V NP_059127.2
XM_017004343.1 1392 Missense Mutation GCG,GTG A458V XP_016859832.1

View Full Product Details