Product Details

SNP ID
rs145653260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:39736754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGCTACAGTCCAGAAGAGTGC[G/T]ACTTCTTATATTTACATATGAACGC
Phenotype
MIM: 611751
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
THUMPD2 PubMed Links

Gene Details

Gene
THUMPD2
Gene Name
THUMP domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321468.1 1628 UTR 3 NP_001308397.1
NM_001321469.1 1628 Missense Mutation NP_001308398.1
NM_001321470.1 1628 Missense Mutation NP_001308399.1
NM_001321474.1 1628 UTR 3 NP_001308403.1
NM_001321475.1 1628 UTR 3 NP_001308404.1
NM_001321477.1 1628 Missense Mutation NP_001308406.1
NM_001321478.1 1628 Missense Mutation NP_001308407.1
NM_001321479.1 1628 Missense Mutation NP_001308408.1
NM_001321480.1 1628 Missense Mutation NP_001308409.1
NM_001321481.1 1628 Missense Mutation NP_001308410.1
NM_025264.4 1628 Missense Mutation NP_079540.2
XM_011533119.2 1628 Missense Mutation XP_011531421.1
XM_017005050.1 1628 Missense Mutation XP_016860539.1
XM_017005051.1 1628 UTR 3 XP_016860540.1
XM_017005052.1 1628 Intron XP_016860541.1
XM_017005053.1 1628 Intron XP_016860542.1
XM_017005054.1 1628 Missense Mutation XP_016860543.1

View Full Product Details