Product Details

SNP ID
rs145908714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223052995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATTTTCTTGATCGGAATCATGCT[A/G]GGCTACATGAAATCCAAGAGGCGGG
Phenotype
MIM: 607775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNE4 PubMed Links

Gene Details

Gene
KCNE4
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080671.3 533 Silent Mutation CTA,CTG L106L NP_542402.3

View Full Product Details