Product Details

SNP ID
rs146028885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112058671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCATTGAAAGATCTTAGACAT[A/G]ATGCTAATCACTATGGCTGCTGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM87B PubMed Links

Gene Details

Gene
TMEM87B
Gene Name
transmembrane protein 87B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032824.2 Intron NP_116213.1
XM_005263827.2 Intron XP_005263884.1
XM_006712807.2 Intron XP_006712870.1
XM_017005121.1 Intron XP_016860610.1

View Full Product Details