Product Details

SNP ID
rs146081742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130342099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGCAGGACCAGCGAATCCAG[C/T]TCCGCTCGCAGGTCAAGCGCCGCCA
Phenotype
MIM: 613734 MIM: 612981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC115 PubMed Links

Gene Details

Gene
CCDC115
Gene Name
coiled-coil domain containing 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321118.1 10 Intron NP_001308047.1
NM_001321119.1 10 UTR 5 NP_001308048.1
NM_032357.3 10 Silent Mutation GAA,GAG E9E NP_115733.2
Gene
IMP4
Gene Name
IMP4 homolog, U3 small nucleolar ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320304.1 10 Intron NP_001307233.1
NM_001320305.1 10 Intron NP_001307234.1
NM_001320306.1 10 Intron NP_001307235.1
NM_001320307.1 10 Intron NP_001307236.1
NM_001320309.1 10 Intron NP_001307238.1
NM_001320310.1 10 Intron NP_001307239.1
NM_001320311.1 10 Intron NP_001307240.1
NM_033416.2 10 Intron NP_219484.1

View Full Product Details