Product Details

SNP ID
rs146432822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:134918877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATGGCGTCGGGCCGTGGAGCTT[C/T]TTCTCGCTGGTTCTTTACTCGGGAA
Phenotype
MIM: 603862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNT2 PubMed Links

Gene Details

Gene
CCNT2
Gene Name
cyclin T2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001241.3 58 Missense Mutation TCT,TTT S8F NP_001232.1
NM_001320748.1 58 UTR 5 NP_001307677.1
NM_001320749.1 58 UTR 5 NP_001307678.1
NM_058241.2 58 Missense Mutation TCT,TTT S8F NP_490595.1
XM_017005227.1 58 Missense Mutation TCT,TTT S8F XP_016860716.1
XM_017005228.1 58 Missense Mutation TCT,TTT S8F XP_016860717.1
XM_017005229.1 58 UTR 5 XP_016860718.1
XM_017005230.1 58 UTR 5 XP_016860719.1
XM_017005231.1 58 UTR 5 XP_016860720.1
XM_017005232.1 58 UTR 5 XP_016860721.1
XM_017005233.1 58 Intron XP_016860722.1
Gene
CCNT2-AS1
Gene Name
CCNT2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details