Product Details

SNP ID
rs146447538
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72132262 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTCAGCATGGCCTCCGTCTCCG[G/T]CAGGATCTCGTTCTGGTTGGAGTCC
Phenotype
MIM: 605207
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP26B1 PubMed Links

Gene Details

Gene
CYP26B1
Gene Name
cytochrome P450 family 26 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277742.1 1307 Missense Mutation ACG,CCG T427P NP_001264671.1
NM_019885.3 1307 Missense Mutation ACG,CCG T502P NP_063938.1
XM_005264433.4 1307 Missense Mutation ACG,CCG T444P XP_005264490.1
XM_011532988.1 1307 Missense Mutation ACG,CCG T311P XP_011531290.1

View Full Product Details