Product Details

SNP ID
rs146570389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151251260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCAGAATGTGGAATTTTTTCAG[C/T]GTGCTTTCATAAGATCCACACTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM43 PubMed Links

Gene Details

Gene
RBM43
Gene Name
RNA binding motif protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198557.2 1325 Silent Mutation NP_940959.1
XM_017004052.1 1325 Silent Mutation XP_016859541.1

View Full Product Details