Product Details

SNP ID
rs146740655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206445339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAATGAAACTGCAGAAAAAAAT[A/T]TGGGAGTCCTGGCAGATGAAGACAA
Phenotype
MIM: 603710
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAM23 PubMed Links

Gene Details

Gene
ADAM23
Gene Name
ADAM metallopeptidase domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003812.3 570 Silent Mutation ATG,TTG M83L NP_003803.1
XM_005246932.3 570 Missense Mutation ATG,TTG M83L XP_005246989.1
XM_011512086.2 570 Missense Mutation ATG,TTG M83L XP_011510388.1

View Full Product Details