Product Details

SNP ID
rs146747408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86442071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGTGCTCACGCTCGGAGAAAG[C/T]TGGCCGGTATTGGTGGGGAGGAGGT
Phenotype
MIM: 611512
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM3A PubMed Links

Gene Details

Gene
KDM3A
Gene Name
lysine demethylase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146688.1 82 Silent Mutation AGC,AGT S8S NP_001140160.1
NM_018433.5 82 Silent Mutation AGC,AGT S8S NP_060903.2
XM_006712051.3 82 UTR 5 XP_006712114.1
XM_017004493.1 82 Silent Mutation AGC,AGT S8S XP_016859982.1
XM_017004494.1 82 UTR 5 XP_016859983.1

View Full Product Details