Product Details

SNP ID
rs147176904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:42444297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCATCTCTCGGTAGCAACGTTT[C/G]AGAGTCAAACCGAGTGTCTGAAGAC
Phenotype
MIM: 606767
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNG3 PubMed Links

Gene Details

Gene
KCNG3
Gene Name
potassium voltage-gated channel modifier subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133329.5 1511 Silent Mutation CTC,CTG L316L NP_579875.1
NM_172344.2 1511 Silent Mutation CTC,CTG L305L NP_758847.1

View Full Product Details