Product Details

SNP ID
rs147613754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:120349381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCTTGTTTGAGCGGGGCGAGC[A/G]GCGACTCAACCTAGACGTGCAGTGT
Phenotype
MIM: 147390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INHBB PubMed Links

Gene Details

Gene
INHBB
Gene Name
inhibin beta B subunit
There are no transcripts associated with this gene.

Gene
LOC105373989
Gene Name
putative cuticle collagen 91
There are no transcripts associated with this gene.

View Full Product Details