Product Details
- SNP ID
-
rs147756793
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88451263 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACGCAGCAAATATGGGATATTCA[C/T]TTCTAAATTTCTAACTAGTTAAGGT
- Phenotype
-
MIM: 612351
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXI3
PubMed Links
Gene Details
- Gene
- FOXI3
- Gene Name
- forkhead box I3
View Full Product Details