Product Details

SNP ID
rs148360771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31173066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCAAAACTGCCTTCAGGGGCC[A/G]AGGCAAAATTGGTTTATTAAAACAG
Phenotype
MIM: 610229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN14 PubMed Links

Gene Details

Gene
CAPN14
Gene Name
calpain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145122.1 3914 UTR 3 NP_001138594.1
NM_001321270.1 3914 UTR 3 NP_001308199.1
XM_011532863.2 3914 UTR 3 XP_011531165.1
XM_011532864.2 3914 UTR 3 XP_011531166.1
XM_011532865.1 3914 UTR 3 XP_011531167.1
XM_011532866.2 3914 UTR 3 XP_011531168.1

View Full Product Details