Product Details

SNP ID
rs148581980
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:42444457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAACACAGAGATGTAATACGGC[A/G]TGATTGCCAGTAAATCAATGATGTT
Phenotype
MIM: 606767
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KCNG3 PubMed Links

Gene Details

Gene
KCNG3
Gene Name
potassium voltage-gated channel modifier subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133329.5 1351 Missense Mutation ACG,ATG T263M NP_579875.1
NM_172344.2 1351 Missense Mutation ACG,ATG T252M NP_758847.1

View Full Product Details