Product Details

SNP ID
rs148640565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227981013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTACACAAGTAATACAGTGAT[A/G]GAAACAAGACAATCTTTTATCTTCC
Phenotype
MIM: 611646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPHKAP PubMed Links

Gene Details

Gene
SPHKAP
Gene Name
SPHK1 interactor, AKAP domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142644.1 6405 UTR 3 NP_001136116.1
NM_030623.3 6405 UTR 3 NP_085126.2
XM_005246870.4 6405 UTR 3 XP_005246927.1
XM_006712777.3 6405 UTR 3 XP_006712840.1
XM_006712778.3 6405 UTR 3 XP_006712841.1
XM_011511925.2 6405 UTR 3 XP_011510227.1

View Full Product Details