Product Details

SNP ID
rs148824183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216860003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATCGGCTCTTGCTCCTCCTCATG[C/T]CATGACTCTTTAATTTGCGGCTGGT
Phenotype
MIM: 190231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928278 PubMed Links

Gene Details

Gene
LOC101928278
Gene Name
uncharacterized LOC101928278
There are no transcripts associated with this gene.

Gene
LOC105373876
Gene Name
uncharacterized LOC105373876
There are no transcripts associated with this gene.

Gene
TNP1
Gene Name
transition protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003284.3 57 Missense Mutation GAC,GGC D11G NP_003275.1

View Full Product Details