Product Details

SNP ID
rs149159317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119680958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTGGAGTTCCAATCTCGTACCA[A/C]CTCTTCCCGGATCCCGTGGTCCAAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFAP221 PubMed Links

Gene Details

Gene
CFAP221
Gene Name
cilia and flagella associated protein 221
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271049.1 237 Intron NP_001257978.1
XM_006712353.3 237 Intron XP_006712416.1
XM_017003550.1 237 Intron XP_016859039.1
XM_017003551.1 237 Intron XP_016859040.1
XM_017003552.1 237 Intron XP_016859041.1
XM_017003553.1 237 Intron XP_016859042.1
XM_017003554.1 237 Intron XP_016859043.1
XM_017003555.1 237 Intron XP_016859044.1
XM_017003556.1 237 Intron XP_016859045.1
XM_017003557.1 237 Intron XP_016859046.1
XM_017003558.1 237 Intron XP_016859047.1
XM_017003559.1 237 Intron XP_016859048.1
Gene
TMEM177
Gene Name
transmembrane protein 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105198.1 237 Missense Mutation CAA,CAC Q35H NP_001098668.1
NM_001105199.1 237 Missense Mutation CAA,CAC Q35H NP_001098669.1
NM_030577.2 237 Missense Mutation CAA,CAC Q35H NP_085054.2

View Full Product Details