Product Details

SNP ID
rs149546776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:203246864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTTCTGGTTTGGCAGGCGCCTC[A/G]TGGTTAGTTTGGGCACTGTTGATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP20A1 PubMed Links

Gene Details

Gene
CYP20A1
Gene Name
cytochrome P450 family 20 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177538.2 854 Missense Mutation ATG,GTG M78V NP_803882.1
XM_005246698.3 854 Intron XP_005246755.1
XM_005246699.3 854 Intron XP_005246756.1
XM_005246701.3 854 Intron XP_005246758.1
XM_006712644.3 854 Missense Mutation ATG,GTG M60V XP_006712707.1
XM_006712645.3 854 Missense Mutation ATG,GTG M78V XP_006712708.1
XM_017004555.1 854 Intron XP_016860044.1

View Full Product Details