Product Details

SNP ID
rs149597972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169735096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGATTTTCTGGATGCATTCAGA[A/T]CATTTTACTTGGATGGATTATTTAC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHL23 PubMed Links

Gene Details

Gene
KLHL23
Gene Name
kelch like family member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144711.5 326 Missense Mutation ACA,TCA T28S NP_653312.2
Gene
PHOSPHO2-KLHL23
Gene Name
PHOSPHO2-KLHL23 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199290.1 326 Missense Mutation ACA,TCA T28S NP_001186219.1

View Full Product Details