Product Details

SNP ID
rs149753087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47521801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCACCGTCGCGGGGGTGGTCAT[A/G]CCGAAACCTGTGGAGACAGGGCAGG
Phenotype
MIM: 607366 MIM: 609309
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCG2040054 PubMed Links

Gene Details

Gene
HCG2040054
Gene Name
uncharacterized LOC644093
There are no transcripts associated with this gene.

Gene
KCNK12
Gene Name
potassium two pore domain channel subfamily K member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022055.1 399 Silent Mutation GGC,GGT G133G NP_071338.1
Gene
MSH2
Gene Name
mutS homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000251.2 399 Intron NP_000242.1
NM_001258281.1 399 Intron NP_001245210.1
XM_005264332.3 399 Intron XP_005264389.2
XM_011532867.1 399 Intron XP_011531169.1

View Full Product Details