Product Details

SNP ID
rs150066286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:132417702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCTCCAGCCGCTGGACCGTGTT[C/G]CAGTCCAGCATCTTCGGCGCCTTCG
Phenotype
MIM: 602886
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR39 PubMed Links

Gene Details

Gene
GPR39
Gene Name
G protein-coupled receptor 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001508.2 1129 Missense Mutation TTC,TTG F220L NP_001499.1
XM_011511021.2 1129 Missense Mutation TTC,TTG F220L XP_011509323.1

View Full Product Details