Product Details

SNP ID
rs150151708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233259823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAAGCCCATTGGTTCTCCACCC[C/T]CTAGCCTCTTGCATCCTTTGGTTTG
Phenotype
MIM: 610767
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG16L1 PubMed Links

Gene Details

Gene
ATG16L1
Gene Name
autophagy related 16 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190266.1 Intron NP_001177195.1
NM_001190267.1 Intron NP_001177196.1
NM_017974.3 Intron NP_060444.3
NM_030803.6 Intron NP_110430.5
NM_198890.2 Intron NP_942593.2
XM_005246082.2 Intron XP_005246139.1
XM_005246084.2 Intron XP_005246141.1
XM_005246086.2 Intron XP_005246143.1
XM_006712608.2 Intron XP_006712671.1

View Full Product Details