Product Details

SNP ID
rs150154167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20000513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAATGATGGGACCCTGTTCTGT[A/C]ATTCACACAAATGTGCTGACACCCA
Phenotype
MIM: 602109
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928222 PubMed Links

Gene Details

Gene
LOC101928222
Gene Name
uncharacterized LOC101928222
There are no transcripts associated with this gene.

Gene
MATN3
Gene Name
matrilin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002381.4 1159 Missense Mutation GAC,TAC D366Y NP_002372.1

View Full Product Details